Learn More
Tocris Bioscience™ CPI 203
BET bromodomain inhibitor; arrests cell cycle at G1 phase
Brand:  Tocris Bioscience™ 5331/10
2754.14 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18758188
  • 3490.00 NOK / 10mg
Explore more special offers
Description
Description
CPI 203 is a BET bromodomain inhibitor. Downregulates Myc expression, causes G1 cell cycle arrest and attenuates cell proliferation in human pancreatic neuroendocrine tumors. Arrests the growth of T cell acute lymphoblastic leukemia cells in vitro (EC50 = 91.2 nM). Also enhances the antitumor effect of rapamycin (Cat. No. 1292) in vitro and attenuates rapamycin induced Akt activation. Also enhances fibroblast reprogramming to hiPSCs at low concentration. Orally bioavailable.
Specifications
Specifications
>98%
CPI 203
CPI 203
10 mg
399.9