Learn More
Bollé Safety Coverall Clear Safety Goggles
Click to view available options
Model:
Coverall Autoclave
Coverall Clean
Unit Size:
Each
Pack of 5
Description
Coverall Autoclave:
- High fogging resistance
- Autoclave compatible product
- Overflow chute for liquids
- PLATINUM coating : anti-scratch & anti-fog
Coverall Clean:
- Can be sterilized using gamma, beta irrdiation or etylene oxide sterilization
- Disposable
- Overflow chute for liquids
- PLATINUM LITE, anti-fog coating
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety Goggles |
Frame Color | White |
Model | Coverall Autoclave |
No. per Pack | 1 Ea. |
Weight (Metric) | 79 g |
Material | Polycarbonate |
Frame Material | TPR |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Videos
-
Bolle Safety Platinum efficiency demonstration
-
Bolle Safety Platinum coating
Documents
Product Information
Specification Sheets
COVERALL CLEAN & AUTOCLAVE
File size: 405.5 kB
Promotions
-
Multiple Manufacturers
Essential Eye Protection
Your vision deserves the best protection.Promo Code: 24152Expires: 2025-07-04
Product Content Correction
Product Title
Bollé Safety Coverall Clear Safety Goggles >