Learn More
Hirschmann™ Counting Chambers
Facilitates quick and easy determination of cell concentration in liquid samples. Hirschmann Instruments™ Counting Chambers are available in a variety of depths to fit a range of systems.
Brand:  Hirschmann™ 8100104
 View more versions of this product
Product Code. 10794873
  • 769.00 NOK / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Description
  • Double net ruling
  • Available bright-lined or no, with or without clamps
  • Includes 2 cover glasses for haemocytometer
  • CE acc. IVD 98/79 EC
Specifications
Specifications
Counting Chambers, Neubauer-improved
1Each
0.1mm
2 Haemocytometer Glass Covers
Product Title
Hirschmann™ Counting Chambers > Depth: 0.1mm; Ruled: Double Net