Learn More
Corning™ Aspirator
Ideal for removal of supernatants - an alternative to glass Pasteur pipets
Brand: Corning™ 4930
3780.46 NOK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Includes: Handpiece, grommet for attachment of accessories, and a single-channel adapter for use with disposable pipetter tips.
Description
The Corning™ Aspirator is a device using vacuum to safely and aseptically dispose of supernatant from a variety of laboratory vessels. The aspirator includes a hand piece to which disposable pipet tips are attached during use, a grommet for accessory attachment and single-channel adapter for disposable pipet tips.
An 8-channel adapter is available for working in 96 well plates, see No. 4931. Refer to the accessories tab below.
Specifications
Autoclavable | |
Aspirator | |
Disposable pipette tip, Polyethylene and Polystyrene aspiration pipette, or Pasteur pipette |
Costar | |
Safe removal/disposal of supernatants from a wide range of laboratory vessels | |
Handpiece, grommet for accessory attachment and single channel adapter for disposable pipette tip |
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30