Learn More
Corning™ Universal Fit Pipet Tips: Bulk Packed
Designed to assure a reliable fit with major brand pipetters
Brand: Corning™ 4845
This item is not returnable.
View return policy
Description
Designed to assure a reliable fit with major brand pipetters
- Beveled orifice helps ensure accurate fluid delivery
- 1–200μL tips are graduated at 10, 50 and 100μL
- 100–1000μL tips are graduated at 250, 500, and 1000μL
- Choice of sterile or nonsterile
- Bulk packed tips are nonsterile and very economical
Specifications
1 to 200 μL | |
Yellow | |
Round | |
Universal Fit | |
Polypropylene |
Non-sterile | |
Bulk | |
1000 Tips/Bag, 10,000 Tips/Cs. | |
Yes |
COR012 LIQUID HANDLING SELECTION GUIDE
File size: 2.5 MB
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Product Content Correction
Product Title
Corning™ Universal Fit Pipet Tips: Bulk Packed > 1-200μL;Yellow; Non-sterile; Bulk packed; 1000 tips/bag, 10000 tips/case