Learn More
Corning™ LSE™ Orbital Shaker Clamps
For holding Erlenmeyer flasks
Brand:  Corning™ 480114
 View more versions of this product
Product Code. 11343815
  • 557.22 NOK / Each
Only null left
Estimated Shipment: 23-10-2025
Description
For use with Corning™ LSE™ Orbital Shaker and Corning™ LSE™ Low Speed Orbital Shaker.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
500 mL Erlenmeyer Flask
Clamp
LSE™ orbital shaker and LSE™ low speed orbital shaker
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Corning™ LSE™ Orbital Shaker Clamps > 500 mL