Learn More
Corning™ Low-Volume 384-Well Black Flat Bottom Polystyrene Non-Treated Microplate
Shop All Corning Products
Click to view available options
Color:
Black
Unit Size:
Pack of 50
Description
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Color | Black |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 0.005 to 0.04 mL |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Well Shape | Flat Bottom (F-well) |
| Show More |