Learn More
Corning™ DeckWorks™ Reload Systems with Standard Tips
Packaged in transparent trays for easy tip identification. Corning™ Deckworks™ Reload Systems with Standard Tips provide convenient and economical reloading of durable hinged racks
Brand: Corning™ 4127
506.22 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Corning DeckWorks reload systems are available in 10 ÎĽL, 200 ÎĽL, 300 ÎĽL, and 1,000 ÎĽL sizes and allow for convenient and economical reloading of durable Corning DeckWorks hinged racks. DeckWorks reload decks are packaged in transparent trays for easy tip identification, requires no transfer devices, saves space, and minimizes packaging waste. Although the pipet tips are made from virgin medical-grade resins, the reload decks and rack bases are manufactured from 100% recycled polypropylene. Nonsterile decks and racks are steam autoclavable.
Corning DeckWorks low binding tips are available in 10 ÎĽL, 200 ÎĽL, and 300 ÎĽL sizes. Proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips is eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins, and/or peptides.
Specifications
1 to 200 ÎĽL | |
Yellow | |
Round | |
Yes | |
Polypropylene |
Non-sterile | |
Autoclavable | |
Reload system | |
DeckWorks |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
Days
72Hours
9Minutes
33Seconds
56New Offer Save Up to 59% -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
31Hours
9Minutes
33Seconds
56