Learn More
Corning™ CrystalEX384-Well Flat Bottom Protein Crystallization Microplate
The only microplate designed for full automation in crystal screening
Brand: Corning™ 3775
This item is not returnable.
View return policy
Description
96-well Microplates
- Features 96 large reservoir (reagent) wells and 96 corresponding protein wells
- Conical bottom protein wells allow improved centering of the protein drop.
- Compatible with manual pipettors and automation
- Novel merged well design provides efficient vapor space for protein crystallization.
- Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
384-well Microplate
- Meets ANSI/SBS standard footprint dimensions for microplates
- Ideal for fully automated crystal screening
- Features 192 reservoir wells and 192 corresponding protein wells
- Flat bottom protein wells are optimized for imaging of crystals.
- Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
Specifications
Standard | |
16 x 24 | |
Advanced Polymer | |
Non-sterile | |
Flat Bottom (F-well) | |
50/Cs. | |
50/Cs. |
Without Lid | |
Clear | |
384 | |
Non-treated | |
192 reservoir wells and 192 corresponding protein wells | |
10/Pk. | |
2.3 mm |
COR001 Lifescience Guid
File size: 7 MB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1419265 | Certificate of Processing | 22/01/2025 | 3775 |
Product Content Correction
Product Title
Corning™ CrystalEX384-Well Flat Bottom Protein Crystallization Microplate > None; 384-well