Learn More
Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™ Product Code.: 15353651
Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™
BIOPHARMA
Product Code. p-2797003
6 x 100 mL, Bottle, Pack of 6
Click to view available options
Quantity:
6 x 100 mL
6 x 500 mL
20 L
6 x 1 L
1 x 100 L
1 x 200 L
Packaging:
Bioprocess Container
Bottle
Unit Size:
100 L
20 L
200 L
Pack of 6
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Product Code. 15353651
Brand: Corning Media Tech 25055CI
Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.
Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).
Order Info
Supplied in a plastic bottle except MT-25-055-LB and -LH, which come in flexible media bags.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Bottle
Quantity 6 x 100 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number31724019 Certificate TypeCertificate of Quality Date22/06/2025 Product Code25-055-CI
Lot Number01325006 Certificate TypeCertificate of Quality Date22/06/2025 Product Code25-055-CI
Lot Number19124018 Certificate TypeCertificate of Quality Date31/05/2025 Product Code25-055-CI
Lot Number16624004 Certificate TypeCertificate of Quality Date25/01/2025 Product Code25-055-CI
Lot Number10024012 Certificate TypeCertificate of Quality Date07/11/2024 Product Code25-055-CI
Product Content Correction
Product Title
Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™ >