Learn More
Corning™ Breathable Sealing Tape for Microplates
Fits 96- and 384-well microplates, polypropylene blocks
Brand:  Corning™ 3345
6862.91 NOK valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Product Code. 10173582
  • 7904.12 NOK / Pack of 500
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Description
  • Easy application and removal for short- and long-term storage
  • Provides tight seal to minimize evaporation and condensation
  • Breathable sealing tape allows gas exchange across the surface
  • Sealing tape 3345 is not transparent
Other Available Options:
  • Universal Optical sealing tape (Cat. No. 6575) is suitable for use between -70°C and 100°C, and is transparent.
  • Aluminum sealing tape (Cat. Nos. 6569 and 6570) is suitable for use between -80°C and 150°C, is not transparent, and is pierceable.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sterile
50/Pk.
Yes
500/Cs.
Breathable Sealing Tape
Documents
Product Information
Product Title
Corning™ Breathable Sealing Tape for Microplates > Breathable