Learn More
Corning™ BioCoat™ Control Insert, PET Membrane
GREENER_CHOICE
Use alongside BD BioCoat™ inserts when studying effects of the ECM component present on the BD BioCoat cell culture insert
Brand:  Corning™ 354578
 View more versions of this product
Product Code. 10274901
  • 3794.44 NOK / Pack of 24
Estimated Shipment: 06-06-2025
Description
Corning BioCoat cell culture inserts are pre-coated with extracellular matrix proteins for applications requiring a protein-coated cell surface, such as cell adhesion, growth, invasion, migration and/or differentiation.
Coatings include Corning Matrigel™ matrix, Fibronectin, Collagen, and Laminin.
Order Info
Supplied in ready-to-use Falcon™ Companion Tissue Culture Plates
TRUSTED_SUSTAINABILITY
Specifications
Specifications
12
PET
Well Plate
2° to 8°C
24/Cs.
24
0.3 cm2
8 μm
2
12/Pk.
BioCoat Control
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Corning™ BioCoat™ Control Insert, PET Membrane > 6.4 mm; 8 μm