Learn More
Corning™ BioCoat™ Collagen I, Rat Tail Culture Dish
BIOPHARMAGREENER_CHOICE
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
Brand:  Corning™ 356450
 View more versions of this product
Product Code. 10499012
  • 4163.57 NOK / Pack of 40
Description
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
Order Info
Bulk quantities available; please contact your Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Polystyrene
Clear
Polystyrene
Collagen Type I
Lid
100 mm
Closure Included
Round
58.1 sq. cm
Round
Cell Culture Dish
40/cs.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number00625010 Certificate TypeCertificate of Quality Date03/06/2025 Catalog Number356450
Lot Number35224007 Certificate TypeCertificate of Quality Date26/04/2025 Catalog Number356450
Lot Number28124022 Certificate TypeCertificate of Quality Date05/02/2025 Catalog Number356450
Lot Number23924018 Certificate TypeCertificate of Quality Date18/01/2025 Catalog Number356450
Lot Number21924019 Certificate TypeCertificate of Quality Date03/12/2024 Catalog Number356450
Promotions
Promotions
Product Content Correction
Product Title
Corning™ BioCoat™ Collagen I, Rat Tail Culture Dish > Cell growth area: 58.1cm2; 100mm; 40/cs.