Learn More
Corning™ 384-Well, Ultra-Low Binding, U-Shaped-Bottom Microplate
Color:Black, Clear Bottom; Sterility:Sterile
Brand: Corning™ 4516
3129.83 NOK valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
With their novel and proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.
- Optically clear round bottom with black opaque microplate body
- Covalent attachment of Ultra-Low Attachment (ULA) surface to reduce cellular adhesion to well surface
- Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization.
- Unique design shields each well to minimize well-to-well cross-talk.
- You can culture and assay spheroids in the same microplate without the need for transfer to a new microplate.
Specifications
Standard | |
0.06 cm2 | |
5/Pk. | |
Spheroid Microplate | |
384 | |
Sterile | |
90 μL | |
Closure | |
1/Pk. |
With Lid | |
16 x 24 | |
Black, Clear Bottom | |
Polystyrene | |
20 to 80 μL | |
Ultra-low Binding | |
Round Bottom (U-well) | |
5/Cs. |