Learn More
Chroma Gesellschaft™ Cork Borers
Efficiently and cleanly bores holes in cork. Chroma Gesellschaft™ Cork Borers have a cutting tube mounted with safety under-flange in shaped handle.
Brand:  Chroma Gesellschaft™ 3400
Product Code. 12853952
  • 749.00 NOK / Each
Only null left
Estimated Shipment: 29-08-2025
Add to basket
Description
Description
  • With rod for clearing bores
  • Manufactured from brass
  • Sizes numbered consecutively from 1 upwards in each set
Specifications
Specifications
Cork Borer
Brass
For boring holes in cork.
4 to 10 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Chroma Gesellschaft™ Cork Borers > Size: 4 to 10mm