missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10464-1-AP-20UL
This item is not returnable.
View return policy
Description
CSN4 is a subunit of the JUN activation-domain binding protein 1 (JAB1), containing signalosome complex (or COP9 complex) which has kinase activity and phosphorylates C-JUN. The signalosome complex may play a role in signal transduction.Specifications
COPS4 | |
Polyclonal | |
Unconjugated | |
COPS4 | |
COPS4, CSN4, SGN4, Signalosome subunit 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26891, 360915, 51138 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88544, Q68FS2, Q9BT78 | |
COPS4 | |
COPS4 Fusion Protein Ag0735 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction