Learn More
Copper(II) nitrate trihydrate, 99%, pure Product Code.: 15253346
compliance-icons compliance-icons compliance-icons
Copper(II) nitrate trihydrate, 99%, pure
Product Code. 15253346 Shop All Thermo Scientific Chemicals Products
5 kg, 5kg
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
Product Code. 15253346
Brand: Thermo Scientific Acros 453960050
Request Bulk or Custom Format
CAS: 10031-43-3 | CuN2O6·3H2O | 241.6 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10031-43-3
Molecular Formula CuN2O6·3H2O
Molecular Weight (g/mol) 241.6
InChI Key SXTLQDJHRPXDSB-UHFFFAOYSA-N
Synonym copper ii nitrate trihydrate, copper nitrate trihydrate, cupric nitrate trihydrate, copper ii nitrate hydrate, copper ii nitrate 3-hydrate, copper 2+ trihydrate dinitronate, copper 2+ trihydrate dinitrate, copper ii nitrate trihydrate-cu puratremShow MoreShow Less
PubChem CID 9837674
IUPAC Name copper;dinitrate;trihydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.[Cu+2]
Specifications
Melting Point 114.0°C
Quantity 5 kg
Assay Percent Range 98.5 to 104% (Iodometry)
Linear Formula Cu(NO3)2·3H2O
Merck Index 15,2633
Solubility Information Solubility in water: 267 g/100 mL. Other solubilities: freely soluble in alcohol, practically insoluble in ethyl acetate
Formula Weight 241.6
Percent Purity 99%
Grade Pure
Chemical Name or Material Copper(II) nitrate trihydrate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0471702 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code453960010,453960050
Lot NumberA0466245 Certificate TypeCertificate of Analysis Date01/10/2024 Product Code453960010,453960050
Lot NumberA0461960 Certificate TypeCertificate of Analysis Date24/04/2024 Product Code453960010,453960050
Lot NumberA0450984 Certificate TypeCertificate of Analysis Date16/04/2024 Product Code453960010,453960050
Lot NumberA0447983 Certificate TypeCertificate of Analysis Date16/04/2024 Product Code453960010,453960050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) nitrate, trihydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Copper(II) nitrate trihydrate, 99%, pure >