missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Nitrate Trihydrate, 95+%, Extra Pure, SLR, Fisher Chemical™

228.00 NOK - 438.00 NOK

Chemical Identifiers

CAS 10031-43-3
Molecular Formula CuN2O6
Molecular Weight (g/mol) 187.55
MDL Number 149671
InChI Key XTVVROIMIGLXTD-UHFFFAOYSA-N
IUPAC Name copper(2+) dinitrate
SMILES [Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11498133
SDS View Documents
Fisher Chemical
C/8080/50
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10796471
SDS View Documents
Fisher Chemical
C/8080/53
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

10031-43-3
187.55
XTVVROIMIGLXTD-UHFFFAOYSA-N
[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
CuN2O6
149671
copper(2+) dinitrate

Specifications

114°C
Blue
Solid
149671
[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
187.55
Glass Jar
10031-43-3
4
CuN2O6
XTVVROIMIGLXTD-UHFFFAOYSA-N
copper(2+) dinitrate
241.6g/mol
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(II) Nitrate Trihydrate, 95+%, Extra Pure, SLR, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.