Learn More
SymLine Continuous Level Control Accessories
Accessories for Continuous Level Control
Brand:  SymLine 117982
392.05 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 14663394
  • 570.00 NOK / Each
Only null left
Estimated Shipment: 04-07-2025
Description
Measuring filling levels continuously - Monitoring non-transparent canisters on a permanent basis - No problem with the continuous level control unit!
Specifications
Specifications
Grounding cable, 1 x clamp, 1 x ring ID 5 mm, L = 1.5 m
1.5 m
1 x clamp, 1 x ring ID 5 mm
Grounding Cable
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SymLine Continuous Level Control Accessories > Type: Grounding cable; 1x clamp, 1x ring ID 5 mm