Learn More
Contec™ Bucket Wringers and Frames
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
Brand: Contec™ 2710
Description
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.- Choose from downpress, flat or curved styles
- Available in polypropylene or stainless steel
- Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
- Select wringer that complements mop
Specifications
Heavy Duty Downpress Mop Wringer | |
Reusable | |
Contec™ Edgeless™ VertiKlean MAX™ or QuickTask™ Mops, 5 gal. (20 L) or 6.5 gal. (25 L) Bucket |
Autoclavable | |
Ideal for cleanrooms and controlled environments | |
Non-sterile |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.