missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Connexin 26 Monoclonal Antibody (CX-1E8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 335800
This item is not returnable.
View return policy
Description
33-5800 was used in the immunofluorescence and western blot analysis to successfully detect Connexin26 in mouse liver sections and homogenate, respectively.
Connexin 26 Monoclonal antibody specifically detects Connexin 26 in Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry (Frozen), Western Blot.Specifications
Connexin 26 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P21994, Q00977 | |
Gjb2 | |
A 13 amino acid synthetic peptide derived from the C-terminus of the mouse Connexin 26 protein. | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG2a κ |
ELISA, Immunohistochemistry (Frozen), Western Blot | |
CX-1E8 | |
Unconjugated | |
Gjb2 | |
AI325222; Cnx26; connexin 26; connexin-26; CX26; CXN-26; DFNA3; DFNA3A; DFNB1; DFNB1A; Gap junction beta-2 protein; gap junction channel protein connexin 26; gap junction membrane channel protein beta 2; gap junction protein beta 2; gap junction protein, beta 2; gap junction protein, beta 2, 26kDa; GJB2; Gjb-2; HID; KID; mutant gap junction protein beta 2; NSRD1; PPK | |
Mouse | |
Protein A | |
RUO | |
14619, 394266 | |
-20°C | |
Liquid |