Learn More
Masterflex™ DB9 Male 9-pin Connector for Creating Your Own Cable
Create your own cable for optimization of your Masterflex™ drive
Brand:  Masterflex™ 07595-45
Product Code. 11847172
  • 1475.00 NOK / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Description
Choose the correct connection for your Masterflex™ pump. Combine these connectors with 20-22 gauge shielded wire to create your own interface cable. Maximum cable length is 1000 ft (300 m).
Specifications
Specifications
DB9 male 9-pin connector for use with drives.