Learn More
SI Analytics™ Connection Cable
SI Analytics™ Connection Cable is for use with the Z890 printer as well as select ProLab meters.
Brand: SI Analytics™ 285203730
1169.88 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with Z890 printer
- Suitable for all ProLab meters except Lab 850
Specifications
Connection Cable | |
Connecting printer Z890 to the ProLab meters (except lab 850) and ProLab-meter family |
Product Content Correction
Product Title
SI Analytics™ Connection Cable > Type: Connecting Cable