Learn More
JG Finneran Associates™ Conical Inserts
Use these inserts for a variety of applications. JG Finneran™ Conical Inserts feature a polymer spring which acts as a shock absorber and protects against breakage if the needle bottoms out.
221.00 NOK - 1120.00 NOK
Specifications
Quantity 100 Pk.
View More Specs
Products 5
Product Code Brand Volume (Metric) Material Price Quantity & Availability  
Product Code Brand Volume (Metric) Material Price Quantity & Availability  
12327019
View Documents
JG Finneran Associates™
4025BS-629
250 ÎĽL Glass
851.00 NOK
Pack of 100
Only null left
Estimated Shipment: 29-07-2025
Add to basket
12337019
missing translation for 'viewDocuments'
JG Finneran Associates™
4025PBS-631
250 ÎĽL Polypropylene
221.00 NOK
Pack of 100
Only null left
Estimated Shipment: 22-07-2025
Add to basket
12377009
missing translation for 'viewDocuments'
JG Finneran Associates™
401PBS-530
100 ÎĽL Polypropylene
302.00 NOK
Pack of 100
Only null left
Estimated Shipment: 22-07-2025
Add to basket
10537352
missing translation for 'viewDocuments'
JG Finneran Associates™
4005BS-625
50 ÎĽL Glass
697.00 NOK
Pack of 100
Only null left
Estimated Shipment: 13-08-2025
Add to basket
10340571
missing translation for 'viewDocuments'
JG Finneran Associates™
401TS-529
100 ÎĽL Glass
1120.00 NOK
Pack of 100
Only null left
Estimated Shipment: 13-08-2025
Add to basket
Description
Description
  • Available preassembled with a polymer bottom spring or patented Top Spring™
  • Polymer spring acts as a shock absorber that protects against breakage if the needle bottoms out
  • Conical design permits complete sample removal
Specifications
Specifications
100 Pk.
Product Title
JG Finneran Associates™ Conical Inserts