Learn More
Lenz Laborglasintrumente Condensers Acc. to Davies, with GL Thread
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
1655.00 NOK - 1805.00 NOK
Product Code | Brand | Length (Metric) | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Standard Taper Joint No | Price | Quantity & Availability | |||||
11546794
|
Lenz Laborglasintrumente
05215314 |
250 mm | NS 29/32 |
1655.00 NOK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
11556794
|
Lenz Laborglasintrumente
05215413 |
400 mm | NS 24/29 |
1805.00 NOK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
11566794
|
Lenz Laborglasintrumente
05215414 |
400 mm | NS 29/32 |
1805.00 NOK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
|||||
Description
- Davies condensers for condensing vapours
- With NS socket and NS cone
- With double sized cooling area
- Water connection: GL 14 threads (with two threaded plastic olives)
- Made of DURAN™ tubing
Specifications
Duran Glass |
Product Title
Lenz Laborglasintrumente Condensers Acc. to Davies, with GL Thread