Learn More
Lenz Laborglasintrumente Condensers acc. to Allihn, with GL Threads
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
839.00 NOK - 952.00 NOK
Specifications
Length (Metric) Jacket 160 mm
View More Specs
Products 2
Product Code Brand Standard Taper Joint No Price Quantity & Availability  
Product Code Brand Standard Taper Joint No Price Quantity & Availability  
10233652
View Documents
Lenz Laborglasintrumente
05223212
NS 19/26
839.00 NOK
Each
Estimated Shipment: 30-07-2025
Add to basket
11943421
View Documents
Lenz Laborglasintrumente
05223213
NS24/29
952.00 NOK
Each
Estimated Shipment: 30-07-2025
Add to basket
Description
Description
  • Allihn condensers acc. to DIN 12581 and in addition to DIN
  • For condensing vapours
  • With NS socket and NS cone
  • Water connection: GL 14 threads (with two threaded plastic olives)
  • Made of DURAN™ tubing
Specifications
Specifications
160 mm
Documents
Product Certifications
Product Content Correction
Product Title
Lenz Laborglasintrumente Condensers acc. to Allihn, with GL Threads