Learn More
Lenz Laborglasintrumente Condensers Acc. to Allihn
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
749.00 NOK - 937.00 NOK
Product Code | Brand | Standard Taper Joint No | No. of Bulbs | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | No. of Bulbs | Price | Quantity & Availability | |||||
11856204
|
Lenz Laborglasintrumente
05223204 |
NS 29/32 | 6 |
761.00 NOK
Each |
Estimated Shipment: 26-08-2025
Log in to see stock. |
|||||
10293951
|
Lenz Laborglasintrumente
05223303 |
NS 24/29 | 5 |
826.00 NOK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
10162091
|
Lenz Laborglasintrumente
05223203 |
NS 24/29 | 6 |
761.00 NOK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
10203911
|
Lenz Laborglasintrumente
05223202 |
NS 19/26 | 6 |
749.00 NOK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
10254051
|
Lenz Laborglasintrumente
05223403 |
NS 24/29 | 8 |
937.00 NOK
Each |
Estimated Shipment: 25-08-2025
Log in to see stock. |
|||||
Description
- Allihn condensers acc. to DIN 12581
- For condensing vapours
- With NS socket and NS cone
- Water connection: glass olives
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) | |
Glass olives |
Product Title
Lenz Laborglasintrumente Condensers Acc. to Allihn