Learn More
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 2mm Thick
For use in preparatory electrophoresis and high-throughput screening.
285.00 NOK - 331.00 NOK
Specifications
For Use With (Equipment) | MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit |
---|---|
Type | Comb |
Quantity | 1 Each |
Thickness (Metric) | 2 mm |
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Wells | Volume (Metric) Sample | Price | Quantity & Availability | |||||
11863363
|
Fisherbrand™
MS15-10-2 |
10 | 90 ÎĽL |
331.00 NOK
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
11803373
|
Fisherbrand™
MS15-10MC-2 |
10 | 59 ÎĽL |
331.00 NOK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
|||||
11833373
|
Fisherbrand™
MS15-1-2 |
1 Prep, 1 Marker | 990 ÎĽL |
331.00 NOK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
|||||
11873373
|
Fisherbrand™
MS15-12-2 |
12 | 81 ÎĽL |
331.00 NOK
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
11813383
|
Fisherbrand™
MS15-14MC-2 |
14 | 59 ÎĽL |
331.00 NOK
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
11853383
|
Fisherbrand™
MS15-16MC-2 |
16 | 54 ÎĽL |
331.00 NOK
Each |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
11893383
|
Fisherbrand™
MS15-18MC-2 |
18 | 23 ÎĽL | N/A | ||||||
11843393
|
Fisherbrand™
MS15-20-2 |
20 | 43 ÎĽL |
331.00 NOK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
11873393
|
Fisherbrand™
MS15-2-2 |
2 Prep, 2 Marker | 450 ÎĽL |
331.00 NOK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
|||||
11813403
|
Fisherbrand™
MS15-28MCSS-2 |
28 | 23 ÎĽL |
331.00 NOK
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
|||||
11853403
|
Fisherbrand™
MS15-30MC-2 |
30 | 25 ÎĽL | N/A | ||||||
11893403
|
Fisherbrand™
MS15-35-2 |
35 | 20 ÎĽL |
331.00 NOK
Each |
Estimated Shipment: 13-08-2025
Log in to see stock. |
|||||
11833413
|
Fisherbrand™
MS15-4-2 |
4 Prep, 2 Marker | 243 ÎĽL |
331.00 NOK
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
|||||
Description
- Recommended for high resolution gels and publication quality data
- Maximise sample volume
- Height adjustable to give user full control over well depth and sample-loading volume
- Rigid back which prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single and multichannel pipettes
- 10, 14, 16, 18, 28 and 30-sample combs are Multichannel pipettor compatible
Specifications
MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit | |
Comb | |
1 Each | |
2 mm |
Product Title
Fisherbrand™ Combs for MultiSUB Midi-Plus Horizontal Gel Electrophoresis Unit, 2mm Thick