Learn More
SCHOTT AG Lighting and Imaging™ Column Adapter for Combi Light Guide
Column Adapter for use with Schott Combi Light Guide 170 402.
Brand:  SCHOTT AG Lighting and Imaging™ 158402
Product Code. 15733032
  • 1905.00 NOK / 1 unit
Estimated Shipment: 13-08-2025
Explore more special offers
Description
  • Positions the Combi light guide (Schott part 170 402) on a 32 mm diameter column.
Specifications
Specifications
Adapter
32 mm
SCHOTT Combi Lightguide (170 402)
1.3 in.
Stereo Microscopy Illumination
Product Title
SCHOTT AG Lighting and Imaging™ Column Adapter for Combi Light Guide > Column Adapter; For Use With: SCHOTT Combi Light Guide (170 402)