Learn More
Lauda™ Colorless Heat Transfer Liquid
9800.00 NOK - 12550.00 NOK
Specifications
Capacity (Metric) 10 L
Type Bath Filler
Quantity 10 L
View More Specs
Products 2
Product Code Brand Ambient Temperature Temperature Range Viscosity Price Quantity & Availability  
Product Code Brand Ambient Temperature Temperature Range Viscosity Price Quantity & Availability  
10656424
View Documents Promotion Details
Lauda™
LZB 221
–50 to 120°C -50°C to +120°C 5mm2/sec.
9800.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 24-10-2024
to see stock.
Add to basket
10244872
View Documents Promotion Details
Lauda™
LZB 216
–20 to 180°C -20°C to +180°C 11mm2/sec.
12550.00 NOK
10 L
View alternative products
Save Up to 
»
Estimated Shipment: 22-10-2024
Specifications
10 L
Bath Filler
10 L