Learn More
Heidolph instruments Collecting Flask
Brand:  Heidolph instruments 514-84000-04
Product Code. 11741689
  • 2220.00 NOK / Each
Estimated Shipment: 10-07-2025
Specifications
Borosilicate Glass
Flask
1000 mL
115/230 VAC
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Heidolph instruments Collecting Flask > Capacity: 1000mL