missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV Monoclonal Antibody (CIV 22), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513437
This item is not returnable.
View return policy
Description
MA5-13437 targets Collagen IV in IHC (P) applications and shows reactivity with Bovine and Human samples. The MA5-13437 immunogen is human glomeruli.
Collagen IV Monoclonal antibody specifically detects Collagen IV in Bovine, Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
Collagen IV | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 15mM sodium azide | |
P02462, P08572, P29400, P53420, Q01955, Q28084, Q29442, Q7SIB2, Q7SIB3 | |
COL4A1, COL4A2, Col4a3, COL4A4, COL4A5 | |
Human glomeruli | |
RUO | |
1282, 1284, 1285, 1286, 1287, 282191, 317711, 407107, 508632, 511602 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin) | |
CIV 22 | |
Unconjugated | |
COL4A2 | |
[a]3(IV); [a]4(IV); alpha 1 type IV collagen; alpha type IV collagen basement membrane; alpha1(IV) collagen; alpha-2 collagen type IV; alpha-3 type IV collagen; alpha3(IV); Arresten; Bru; BSVD; CA44; Canstatin; Col4a1; Col4a-1; COL4A1 NC1 domain; Col4a2; Col4a-2; COL4A3; col4a4; Collagen 4; Collagen alpha-1(IV) chain; collagen alpha-2(IV) chain; Collagen alpha-3(IV) chain; collagen alpha-4(IV) chain; collagen IV, alpha-1 polypeptide; collagen IV, alpha-3 polypeptide; Collagen IV, alpha-4 polypeptide; collagen of basement membrane, alpha-1 chain; collagen of basement membrane, alpha-4 chain; collagen type IV alpha 1 chain; collagen type IV alpha 2; collagen type IV alpha 2 chain; collagen type IV alpha 3 chain; collagen type IV alpha 4 chain; collagen type IV alpha3 chain; collagen, type IV, alpha 1; collagen, type IV, alpha 2; collagen, type IV, alpha 3; collagen, type IV, alpha 3 (Goodpasture antigen); collagen, type IV, alpha 4; Del(8)44H; Del(8)Bru44H; E130010M05Rik; Goodpasture antigen; ICH; LOW QUALITY PROTEIN: collagen alpha-1(IV) chain; LOW QUALITY PROTEIN: collagen alpha-3(IV) chain; LOW QUALITY PROTEIN: collagen alpha-4(IV) chain; POREN2; procollagen alpha1(IV); procollagen, type IV, alpha 1; procollagen, type IV, alpha 2; procollagen, type IV, alpha 3; procollagen, type IV, alpha 4; RATOR; Raw; retinal anterior wiring; Svc; tumstatin; Type IV Collagen | |
Mouse | |
500 μL | |
Primary | |
Bovine, Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction