Learn More
Conductivity Solution, Oakton™ Product Code.: 11765509
Conductivity Solution, Oakton™
Product Code. 11765509 Shop All Cole-Parmer Products
1.5 mS (1500 μS), Each
Click to view available options
Conductivity:
0.023 mS (23 μS)
0.447 mS (447 μS)
1.413 mS (1413 μS)
1.5 mS (1500 μS)
Unit Size:
Each
Product Code. 11765509
Brand: Cole Parmer 0065315
Use this conductivity solution for accurate test results in the field or lab. Oakton™ Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
Use this conductivity solution for accurate test results in the field or lab. OaktonTM Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
  • Features temperature corrections printed on the bottle
  • Certified values listed in the tables and are referenced to 25°C
  • Certificates of analysis available separately
Specifications
Chemical Name or Material Cole-Parmer Conductivity Standards
Packaging Bottle
Conductivity 1.5 mS (1500 μS)
Technique Calibration of Conductivity Instrument
Color Clear
pH 4.7 to 7.8
Boiling Point 100°C (212°F)
For Use With (Equipment) Conductivity Meter
Solubility Soluble
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Show More Show Less
Hazard Statement
  • EUH210-Safety data sheet available on request.
Product Content Correction
Product Title
Conductivity Solution, Oakton™ >