Learn More
Brady™ Codesoft Lite Online Software
Codesoft Lite allows you to create professional labels for any project, including barcoding.
Brand: Brady™ 362854
Description
The Codesoft Lite online software is for use with Brady™ Label Printers. Compatible with BBP™11 Label Printer, BBP™12 Industrial Label Printer, BBP™72 Double-sided Sleeve Printer, BradyPrinter PR Plus Series Label Printer, IP™ Series Printer, PAM3000 Print and Apply Machine, PAM3600 Print and Apply Machine, THT 76 mm Core Printers, and TLS-PC LINK™ Thermal Labeling System.
This enterprise-level label design software allows users to design and print custom labels for product identification and tracking. Codesoft software is the professional solution for the most complex barcode labeling projects. This web-based software provides a range of barcode label design features for any project, from easy label designs to general barcoding.
Requires:
- 32 or 64 bit Windows operating systems: 7, XP, Server 2003 or 2008 or Vista
- Permanent internet connection
- 1 GB of free RAM
- Hard drive with at least 1 GB free disk space (depending on the installation options)
- CPU: 2 Ghz minimum
- Video card: DirectX 9 minimum
Specifications
Brady Label Printers | |
Web Based Software | |
Asset Tracking, Barcode Labeling, Inventory and Inspection Labeling | |
CS LT SWON 1 SMA |
Arabic, Czech, Danish, Dutch, English, Finnish, French, German, Hungarian, Italian, Norwegian, Polish, Portuguese, Romanian, Russian, Spanish, Swedish, Turkish | |
Windows™ 7 | |
Service Contract: 1 Year |