Learn More
COD Standard, 500mg/L, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
Concentration | 500 mg/L |
Quantity | 500 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
COD50024G1 | Certificate of Analysis | 29/01/2025 | COD500 |
COD50024G1 | Certificate of Analysis | 23/07/2024 | COD500 |
COD50023G1 | Certificate of Analysis | 27/09/2023 | COD500 |
COD50023C1 | Certificate of Analysis | 19/04/2023 | COD500 |
COD50021C1 | Certificate of Analysis | 23/11/2021 | COD500 |
Product Content Correction
Product Title
COD Standard, 500mg/L, Reagecon™ >