Learn More
COD Standard, 1300mg/L, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
Concentration | 1300 mg/L |
Quantity | 500 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
COD130025B1 | Certificate of Analysis | 24/06/2025 | COD1300 |
COD130023G1 | Certificate of Analysis | 02/07/2024 | COD1300 |
COD130022C1 | Certificate of Analysis | 15/03/2023 | COD1300 |
COD130022F1 | Certificate of Analysis | 16/11/2022 | COD1300 |
Product Content Correction
Product Title
COD Standard, 1300mg/L, Reagecon™ >