Learn More
COD Standard, 100mg/L, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
Concentration | 100 mg/L |
Quantity | 500 mL |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number | Certificate Type | Date | Product Code |
---|---|---|---|
COD10024F1 | Certificate of Analysis | 15/04/2025 | COD100 |
COD10023K1 | Certificate of Analysis | 15/03/2024 | COD100 |
COD10022G1 | Certificate of Analysis | 08/12/2022 | COD100 |
COD10022C1 | Certificate of Analysis | 21/07/2022 | COD100 |
Product Content Correction
Product Title
COD Standard, 100mg/L, Reagecon™ >