Learn More
Bollé Safety Cobra Clear Safety Glasses Product Code.: 15612617
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
Bollé Safety Cobra Clear Safety Glasses
Product Code. 15612617
COBTPRPSI
Click to view available options
Model:
COBFSPSI
COBFTPSI
COBHDPI
COBTPRPSI
Product Code. 15612617
Brand: Bollé Safety COBTPRPSI
Estimated Shipment: 20-05-2025
Cobra Safety Glasses come with anti-scratch/anti-fog coatings.
  • Clear
  • Anti-scratch coating
  • Anti-fog coating
  • Easy to clean
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN166, EN170/2C-1.2, Mechanical risk B/T, Chemical risk 3/4/5, Anti-fog N, Scratch-resistant K, Optical class 1, CE
Lens Type Chemical-resistant
Lens Coating Anti-fog, Impact-resistant, Scratch-resistant, UV-treated
Lens Tint Clear
Product Type Safety Glasses
For Use With (Application) Indoor, protection, continuous wear
Includes Microfiber pouch
Model COBTPRPSI
Weight (Metric) 34 g
Lens Material Polycarbonate
  • Bollé Safety - Platinum efficiency demonstration
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Bolle Safety Platinum efficiency demonstration
    • Bollé Safety - Cobra
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Bolle Safety Cobra
      • Bollé Safety - Platinum coating
        Video Player is loading.
        Current Time 0:00
        Duration 0:00
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 0:00
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
          Bolle Safety Platinum coating
        Specification Sheets
        • Multiple Manufacturers
          Save up to 60% on gloves, apparel, and more
          Promo Code: 24131
          Expires2025-06-08
          New Offer
          Shop Now
        Product Content Correction
        Product Title
        Bollé Safety Cobra Clear Safety Glasses >