Learn More
Bollé Safety Cobra Clear Safety Glasses Product Code.: 12649150
Add to basket
Bollé Safety Cobra Clear Safety Glasses
Product Code. 12649150
COBFSPSI
Click to view available options
Model:
COBFSPSI
COBFTPSI
COBHDPI
COBTPRPSI
Product Code. 12649150
Brand: Bollé Safety COBFSPSI
Add to basket
Add to basket
Cobra Safety Glasses come with anti-scratch/anti-fog coatings.
  • Clear
  • Anti-scratch coating
  • Anti-fog coating
  • Easy to clean
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Safety Glasses
Model COBFSPSI
  • Bollé Safety - Platinum efficiency demonstration
    Video Player is loading.
    Current Time 0:00
    Duration 3:09
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 3:09
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Bolle Safety Platinum efficiency demonstration
  • Bollé Safety - Cobra
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Bolle Safety Cobra
    • Bollé Safety - Platinum coating
      Video Player is loading.
      Current Time 0:00
      Duration 1:06
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 1:06
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Bolle Safety Platinum coating
    Specification Sheets
    • Multiple Manufacturers
      Save up to 60% on gloves, apparel, and more
      Promo Code: 24131
      Expires2025-06-08
      New Offer
      Shop Now
    Product Content Correction
    Product Title
    Bollé Safety Cobra Clear Safety Glasses >