Learn More
Cobalt Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™ Product Code.: 10774593
Cobalt Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
Product Code. 10774593 Shop All Fisher Chemical Products
100 mL
Click to view available options
Quantity:
100 mL
500 mL
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10774593
Brand: Fisher Chemical J/8020/05
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Chemical Identifiers
MDL Number 10935
Specifications
Packaging HDPE plastic bottle
Quantity 100 mL
Physical Form Liquid
Color Colorless
pH 1
compliance-icons
Product Identifier
  • Cobalt solution 1000 ppm
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
  • EUH208-Contains A§. May produce an allergic reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction
Product Title
Cobalt Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™ >