Learn More
Buerkle™ Course Thread Adapter for Ball Valve
With inner thread of 3/4 in.
Brand: Buerkle™ 0600-0140
124.62 NOK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Course thread
- Tri-Sure
- 2 in. outer thread
Specifications
Thread Adapter | |
Polypropylene | |
Ball Valve | |
3/4 in. Internal, 2 in. Tri-Sure™ coarse thread |