missing translation for 'onlineSavingsMsg'
Learn More

CNR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 29708-1-AP-150UL

 View more versions of this product

Product Code. 17659686

  • 4593.76 NOK / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CNR1 Polyclonal antibody apecifically detetects CNR1 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

CNR1
Polyclonal
Unconjugated
BC074812
Rabbit
Antigen affinity purification
Primary
Human, Mouse, Rat
Liquid
Western Blot, ELISA
450 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
CNR1
CNR1 fusion protein Ag31838
150 μL
1268
Store at -20°C. Stable for one year after shipment.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers