missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNKR2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
4250.00 NOK
Specifications
Antigen | CNKR2 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
CNKR2 Polyclonal specifically detects CNKR2 in Human samples. It is validated for Western Blot.Specifications
CNKR2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
CNK homolog protein 2, CNK2MAGUIN, connector enhancer of kinase suppressor of Ras 2, Connector enhancer of KSR 2, KIAA0902KSR2connector enhancer of KSR2, membrane-associated guanylate kinase-interacting protein | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human CNKR2 (NP_055742). Peptide sequence EEEEEEGEAAGENIGEKSESREEKLGDSLQDLYRALEQASLSPLGEHRIS | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
22866 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title