Learn More
CLTC Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26523-1-AP-20UL
 View more versions of this product
Product Code. 16804965
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
The CLTC antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CLTC. This antibody recognizes human, pig, mouse, rat antigen. The CLTC antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
Specifications
CLTC
Polyclonal
Unconjugated
20 μL
1213
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence
300 μg/mL
Rabbit
Primary
Human, Mouse, Rat, Pig