Learn More
Tenak™ Clip-on Label Holder
Clip-on Label Holder for easy organizing of a freezer.
148.00 NOK - 12605.00 NOK
Specifications
For Use With (Equipment) | Racks for both upright freezers and chest freezers |
---|---|
Type | Clip-on Label Holder |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15201014
|
Tenak™
TE76110 |
10/Pk. |
1360.00 NOK
Pack of 10 |
Estimated Shipment: 25-03-2025
Log in to see stock. |
|||||
15211014
|
Tenak™
TE76120 |
20/Pk. |
12605.00 NOK
Pack of 100 |
Estimated Shipment: 25-03-2025
Log in to see stock. |
|||||
15291004
|
Tenak™
TE76100 |
1 Ea. |
148.00 NOK
Each |
Estimated Shipment: 25-03-2025
Log in to see stock. |
|||||
Description
- Can be used for racks for both upright freezers and chest freezers
- Universal and can be used for most commonly sold racks of both stainless steel and aluminum with a material thickness of up to 1.5 mm
- Has a handel lifter which makes it easier to grab the rack handel (works with TENAK racks)
- Labels are not included
- Easy to install - Simply just clip it onto the rack and a barb secures it
Specifications
Racks for both upright freezers and chest freezers | |
Clip-on Label Holder |