Learn More
CleanSpace™ Technology CleanSpace™ HALO Neck Support
Plastic neck support is for use with the CleanSpace HALO PAPR units.
Brand: CleanSpace™ Technology CS3009
View more versions of this product
This item is not returnable.
View return policy
Description
- Used with CleanSpace respirators: light weight, no hoses or belts
- Designed for comfort over long periods
- Allows sweating and breathability
- Easy to wash and quick drying
- Designed for long wear in harsh environments
- Easy and quick replacement
Specifications
Reusable | |
Neck Support | |
CS3000 PAPR System |
ABS Plastic | |
Small | |
CS3009 |
Product Content Correction
Product Title
CleanSpace™ Technology CleanSpace™ HALO Neck Support > Small