Learn More
CleanSpace™ HALO Half Mask for PAPR Assembly
Shop All CleanSpace Technology Products
Click to view available options
Size:
Small
Unit Size:
Each
Description
- Used with the revolutionary CleanSpace HALO: light weight, no hoses or belts
- Half masks available in 3 sizes to facilitate a good seal and a comfortable fit
- Materials: Soft silicone cushion, nylon clips and polycarbonate (exhalation valve)
- Mask Fit Test adaptor available for TSI Portacount™ fit testing
- Easy removal
- Compatible with most cleaning and disinfection protocols
- Transparent and facilitates communication
Specifications
Specifications
Certifications/Compliance | AS/NZS1716: 2012 |
Certifications/Compliance (Europe) | EN 12942 |
Product Type | PAPR Half Mask |
Size | Small |
Facepiece/Headtop Model | Half mask only |
For Use With (Application) | Primary healthcare facilities, pharmaceutical production, laboratories, animal research facilities and emergency/disaster responders |
Intrinsic Safety (IS) Rating | No |
Model | CS3003 |
If you require intrinsically safe equipment, please consult your site Intrinsically Safe approvals. These Half Masks are NOT certified intrinsically safe.
Product Content Correction
Product Title
CleanSpace™ HALO Half Mask for PAPR Assembly >