Learn More
Scotch-Brite™ Clean & Shine Floor Pad
This 2 in 1 daily cleaning pad for low speed scrubbers gradually increases shine with repeated use
Brand:  Scotch-Brite™ 7100148013
 View more versions of this product
Product Code. 17188760
  • 2340.00 NOK / Pack of 5
Only null left
Estimated Shipment: 28-07-2025
Description
  • Cleans and shines in the same step, using only a low-speed scrubber
  • Save costs and labor by reducing or eliminating the need to burnish
  • Removes black marks 3 times faster than traditional cleaning pads
  • Two-sided pad lasts longer than traditional cleaning pads
  • Effective on most coated and uncoated hard floors including VCT, vinyl, rubber, marble, stone, terrazzo, granite, and concrete with only water or neutral cleaner
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cleaning Pads
380 mm
Concrete, Granite, Industrial Floors, Linoleum, Marble, Stoneware, Terrazzo, VCT, and Vinyl Flooring
Brown
Cleaning and Polishing Floors
Round
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Scotch-Brite™ Clean & Shine Floor Pad > 380 mm