Learn More
Cytiva Classical Powdered Ham's Nutrient Mixture F10 - F12
Brand: Cytiva SH30010.03
This item is not returnable.
View return policy
Description
Support cell culture growth and biomanufacturing processes with powdered HyClone™ Ham's F10-F12 Nutrient Mixture, manufactured using ISO 9001:2000-certified processes.
Specifications
Powder | |
Non-sterile | |
No Sodium Bicarbonate | |
cGMP and ISO 9001:2000 | |
Ham's F12, with L-Glutamine |
Ham's F12, with L-glutamine | |
Glutamine | |
1 x 10 L | |
2° to 8°C, away from light |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Product Content Correction
Product Title
Cytiva Classical Powdered Ham's Nutrient Mixture F10 - F12 > 1 x 10L