Learn More
Abeba™ Classic 31000 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
918.00 NOK - 949.00 NOK
Specifications
Certifications/Compliance | CE EN ISO 20345:2011 SB SRC |
---|---|
Color | White |
Liner Material | Microfiber |
Material | Leather |
Sole Features | Slip-resistant |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15465684
|
Abeba™
31000/36 |
36 |
918.00 NOK
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15485684
|
Abeba™
31000/37 |
37 |
918.00 NOK
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15495684
|
Abeba™
31000/38 |
38 |
918.00 NOK
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15405694
|
Abeba™
31000/39 |
39 |
918.00 NOK
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
15485694
|
Abeba™
31000/40 |
40 |
949.00 NOK
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
15425704
|
Abeba™
31000/41 |
41 |
949.00 NOK
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
15445704
|
Abeba™
31000/42 |
42 |
949.00 NOK
1 pair |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
15455704
|
Abeba™
31000/43 |
43 |
918.00 NOK
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15475704
|
Abeba™
31000/44 |
44 |
918.00 NOK
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15485704
|
Abeba™
31000/45 |
45 |
918.00 NOK
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
15405714
|
Abeba™
31000/46 |
46 |
949.00 NOK
1 pair |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||||
Description
- CE EN ISO 20345:2011 SB SRC
- Grained leather
- White
- Steel toe cap
- Slip resistance PU outsole sole
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Lining microfiber, respiration absorbing
- Adjustable instep area
- Buckle fastening
- Heel patch microfiber
Specifications
CE EN ISO 20345:2011 SB SRC | |
Microfiber | |
Slip-resistant | |
31000 | |
Animal fats, oil and fuel |
White | |
Leather | |
Polyurethane | |
Steel |